Human IL-4 R alpha, Fc Tag (ILR-H5253) is expressed from human 293 cells (HEK293). It contains AA Met 26 - His 232 (Accession P2439 4-1). 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Human IL-4 R alpha, Fc Tag (ILR-H5253) is expressed from human 293 cells (HEK293). It contains AA Met 26 - His 232 (Accession P2439 4-1). 1 * 100 G
PRODUKT
Erweitert / Kompakt
Human IL-4 R alpha, Fc Tag (ILR-H5253) is expressed from human 293 cells (HEK293). It contains AA Met 26 - His 232 (Accession P2439 4-1). 1 * 100 µG ACRBILR-H5253100UG 1 * 100 µG
Hersteller-Artikelnummer:: ILR-H5253100UG
Lokale Artikelnummer:: ACRBILR-H5253100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder