Human ITGA9ITGB1 Heterodimer Protein, His TagTag Free (IT1-H52W4) is expressed from human 293 cells (HEK293). It contains AA Tyr 30 - Trp 981 (ITGA9) Gln 21 - Asp 728 (ITGB1) (Accession Q13797-1 (ITGA9) P05556-1 (ITGB1)). 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Human ITGA9ITGB1 Heterodimer Protein, His TagTag Free (IT1-H52W4) is expressed from human 293 cells (HEK293). It contains AA Tyr 30 - Trp 981 (ITGA9) Gln 21 - Asp 728 (ITGB1) (Accession Q13797-1 (ITGA9) P05556-1 (ITGB1)). 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Human ITGA9ITGB1 Heterodimer Protein, His TagTag Free (IT1-H52W4) is expressed from human 293 cells (HEK293). It contains AA Tyr 30 - Trp 981 (ITGA9) Gln 21 - Asp 728 (ITGB1) (Accession Q13797-1 (ITGA9) P05556-1 (ITGB1)). 1 * 1 mg ACRBIT1-H52W4-1MG 1 * 1 mg
Hersteller-Artikelnummer:: IT1-H52W4-1MG
Lokale Artikelnummer:: ACRBIT1-H52W4-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder