Biotinylated Human ITGAVITGB1 Heterodimer Protein, His,AvitagTag Free (IT1-H82W6) is expressed from human 293 cells (HEK293). It con tains AA Phe 31 - Val 992 (ITGAV) Gln 21 - Asp 728 (ITGB1) (Accession NP002201.1 (ITGAV) NP002202.2 (ITGB1)). 1 * 25 G
Lieferant: ACROBIOSYSTEMS
Biotinylated Human ITGAVITGB1 Heterodimer Protein, His,AvitagTag Free (IT1-H82W6) is expressed from human 293 cells (HEK293). It con tains AA Phe 31 - Val 992 (ITGAV) Gln 21 - Asp 728 (ITGB1) (Accession NP002201.1 (ITGAV) NP002202.2 (ITGB1)). 1 * 25 G
PRODUKT
Erweitert / Kompakt
Biotinylated Human ITGAVITGB1 Heterodimer Protein, His,AvitagTag Free (IT1-H82W6) is expressed from human 293 cells (HEK293). It con tains AA Phe 31 - Val 992 (ITGAV) Gln 21 - Asp 728 (ITGB1) (Accession NP002201.1 (ITGAV) NP002202.2 (ITGB1)). 1 * 25 µG ACRBIT1-H82W6-25UG 1 * 25 µG
Hersteller-Artikelnummer:: IT1-H82W6-25UG
Lokale Artikelnummer:: ACRBIT1-H82W6-25UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder