Biotinylated Human LAG-3, Fc,Avitag (LA3-H82Fb) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Leu 450 (Accessi on P18627-1). 1 * 25 G
Lieferant: ACROBIOSYSTEMS
Biotinylated Human LAG-3, Fc,Avitag (LA3-H82Fb) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Leu 450 (Accessi on P18627-1). 1 * 25 G
PRODUKT
Erweitert / Kompakt
Biotinylated Human LAG-3, Fc,Avitag (LA3-H82Fb) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Leu 450 (Accessi on P18627-1). 1 * 25 µG ACRBLA3-H82FB-25UG 1 * 25 µG
Hersteller-Artikelnummer:: LA3-H82FB-25UG
Lokale Artikelnummer:: ACRBLA3-H82FB-25UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder