Biotinylated Human LDL R, His,Avitag (LDR-H82E7) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Arg 788 (Access ion P01130-1). 1 * 200 G
Lieferant: ACROBIOSYSTEMS
Biotinylated Human LDL R, His,Avitag (LDR-H82E7) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Arg 788 (Access ion P01130-1). 1 * 200 G
PRODUKT
Erweitert / Kompakt
Biotinylated Human LDL R, His,Avitag (LDR-H82E7) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Arg 788 (Access ion P01130-1). 1 * 200 µG ACRBLDR-H82E7200UG 1 * 200 µG
Hersteller-Artikelnummer:: LDR-H82E7200UG
Lokale Artikelnummer:: ACRBLDR-H82E7200UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder