Mouse LDL R, His Tag (LDR-M52H8) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Arg 790 (Accession NP034830). 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Mouse LDL R, His Tag (LDR-M52H8) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Arg 790 (Accession NP034830). 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Mouse LDL R, His Tag (LDR-M52H8) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Arg 790 (Accession NP034830). 1 * 1 mg ACRBLDR-M52H8-1MG 1 * 1 mg
Hersteller-Artikelnummer:: LDR-M52H8-1MG
Lokale Artikelnummer:: ACRBLDR-M52H8-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder