Human CD58, Fc Tag (LF3-H5256) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Arg 215 (Accession AAH05930). 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Human CD58, Fc Tag (LF3-H5256) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Arg 215 (Accession AAH05930). 1 * 100 G
PRODUKT
Erweitert / Kompakt
Human CD58, Fc Tag (LF3-H5256) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Arg 215 (Accession AAH05930). 1 * 100 µG ACRBLF3-H5256100UG 1 * 100 µG
Hersteller-Artikelnummer:: LF3-H5256100UG
Lokale Artikelnummer:: ACRBLF3-H5256100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder