ActiveMax Human LIF, Tag Free (LIF-H521b) is expressed from human 293 cells (HEK293). It contains AA Ser 23 - Phe 202 (Accession A AH69540.1). 1 * 20 G
Lieferant: ACROBIOSYSTEMS
ActiveMax Human LIF, Tag Free (LIF-H521b) is expressed from human 293 cells (HEK293). It contains AA Ser 23 - Phe 202 (Accession A AH69540.1). 1 * 20 G
PRODUKT
Erweitert / Kompakt
ActiveMax® Human LIF, Tag Free (LIF-H521b) is expressed from human 293 cells (HEK293). It contains AA Ser 23 - Phe 202 (Accession A AH69540.1). 1 * 20 µG ACRBLIF-H521B-20UG 1 * 20 µG
Hersteller-Artikelnummer:: LIF-H521B-20UG
Lokale Artikelnummer:: ACRBLIF-H521B-20UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder