Recombinat Protein,Biotinylated Human LOXL2 Protein, His,Avitag,Biotinylated Human LOXL2, His,Avitag (LO2-H82E3) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Gln 774 (Accession Q9Y4K0-1).This protein carries a polyhistidine t 1 * 25 G
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Biotinylated Human LOXL2 Protein, His,Avitag,Biotinylated Human LOXL2, His,Avitag (LO2-H82E3) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Gln 774 (Accession Q9Y4K0-1).This protein carries a polyhistidine t 1 * 25 G
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Biotinylated Human LOXL2 Protein, His,Avitag™,Biotinylated Human LOXL2, His,Avitag (LO2-H82E3) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Gln 774 (Accession Q9Y4K0-1).This protein carries a polyhistidine t 1 * 25 µG ACRBLO2-H82E3-25UG 1 * 25 µG
Hersteller-Artikelnummer:: LO2-H82E3-25UG
Lokale Artikelnummer:: ACRBLO2-H82E3-25UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder