Human LRP-5, Mouse IgG2a Fc Tag (LR5-H5254) is expressed from human 293 cells (HEK293). It contains AA Glu 644 - Gln 1263 (Accession O75197-1). 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Human LRP-5, Mouse IgG2a Fc Tag (LR5-H5254) is expressed from human 293 cells (HEK293). It contains AA Glu 644 - Gln 1263 (Accession O75197-1). 1 * 100 G
PRODUKT
Erweitert / Kompakt
Human LRP-5, Mouse IgG2a Fc Tag (LR5-H5254) is expressed from human 293 cells (HEK293). It contains AA Glu 644 - Gln 1263 (Accession O75197-1 ). 1 * 100 µG ACRBLR5-H5254100UG 1 * 100 µG
Hersteller-Artikelnummer:: LR5-H5254100UG
Lokale Artikelnummer:: ACRBLR5-H5254100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder