Human MICA, His Tag (MIA-H5221) is expressed from human 293 cells (HEK293). It contains AA Glu 24 - Gln 308 (Accession AAH16929.1) . 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Human MICA, His Tag (MIA-H5221) is expressed from human 293 cells (HEK293). It contains AA Glu 24 - Gln 308 (Accession AAH16929.1) . 1 * 100 G
PRODUKT
Erweitert / Kompakt
Human MICA, His Tag (MIA-H5221) is expressed from human 293 cells (HEK293). It contains AA Glu 24 - Gln 308 (Accession AAH16929.1) . 1 * 100 µG ACRBMIA-H5221100UG 1 * 100 µG
Hersteller-Artikelnummer:: MIA-H5221100UG
Lokale Artikelnummer:: ACRBMIA-H5221100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder