Recombinat Protein,Human MMP-9 Protein, His Tag,Human MMP-9, His Tag (MM9-H5221) is expressed from human 293 cells (HEK293). It cont ains AA Ala 20 - Pro 469 (Accession AAH06093).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human MMP-9 Protein, His Tag,Human MMP-9, His Tag (MM9-H5221) is expressed from human 293 cells (HEK293). It cont ains AA Ala 20 - Pro 469 (Accession AAH06093).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human MMP-9 Protein, His Tag,Human MMP-9, His Tag (MM9-H5221) is expressed from human 293 cells (HEK293). It cont ains AA Ala 20 - Pro 469 (Accession AAH06093).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 1 mg ACRBMM9-H5221-1MG 1 * 1 mg
Hersteller-Artikelnummer:: MM9-H5221-1MG
Lokale Artikelnummer:: ACRBMM9-H5221-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder