Recombinat Protein,Human Mucin-1 MUC-1 Protein, Fc Tag,Human Mucin-1, Fc Tag (MU1-H5252) is expressed from human 293 cells (HEK293) . It contains AA Ser 33 - Gly 167 (Accession AAI20976).This protein carries a human IgG1 Fc tag at the C-terminus. Th 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human Mucin-1 MUC-1 Protein, Fc Tag,Human Mucin-1, Fc Tag (MU1-H5252) is expressed from human 293 cells (HEK293) . It contains AA Ser 33 - Gly 167 (Accession AAI20976).This protein carries a human IgG1 Fc tag at the C-terminus. Th 1 * 100 G
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human Mucin-1 MUC-1 Protein, Fc Tag,Human Mucin-1, Fc Tag (MU1-H5252) is expressed from human 293 cells (HEK293) . It contains AA Ser 33 - Gly 167 (Accession AAI20976).This protein carries a human IgG1 Fc tag at the C-terminus. Th 1 * 100 µG ACRBMU1-H5252100UG 1 * 100 µG
Hersteller-Artikelnummer:: MU1-H5252100UG
Lokale Artikelnummer:: ACRBMU1-H5252100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder