Recombinat Protein,Human NKG2A CD159a Protein, His Tag,Human NKG2A, His Tag (NKA-H5244) is expressed from human 293 cells (HEK293). It contains AA Pro 94 - Leu 233 (Accession P26715-1).This protein carries a polyhistidine tag at the N-terminus. The 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human NKG2A CD159a Protein, His Tag,Human NKG2A, His Tag (NKA-H5244) is expressed from human 293 cells (HEK293). It contains AA Pro 94 - Leu 233 (Accession P26715-1).This protein carries a polyhistidine tag at the N-terminus. The 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human NKG2A CD159a Protein, His Tag,Human NKG2A, His Tag (NKA-H5244) is expressed from human 293 cells (HEK293). It contains AA Pro 94 - Leu 233 (Accession P26715-1).This protein carries a polyhistidine tag at the N-terminus. The 1 * 1 mg ACRBNKA-H5244-1MG 1 * 1 mg
Hersteller-Artikelnummer:: NKA-H5244-1MG
Lokale Artikelnummer:: ACRBNKA-H5244-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder