Human PD-1, His Tag, low endotoxin (HPLC-verified) (PD1-H522a) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - G ln 167 (Accession NP005009.2). 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Human PD-1, His Tag, low endotoxin (HPLC-verified) (PD1-H522a) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - G ln 167 (Accession NP005009.2). 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Human PD-1, His Tag, low endotoxin (HPLC-verified) (PD1-H522a) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - G ln 167 (Accession NP005009.2). 1 * 1 mg ACRBPD1-H522A-1MG 1 * 1 mg
Hersteller-Artikelnummer:: PD1-H522A-1MG
Lokale Artikelnummer:: ACRBPD1-H522A-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder