Human PD-L2, Mouse IgG1 Fc Tag (PD2-H52A5) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 219 (Accession A AI13679). 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Human PD-L2, Mouse IgG1 Fc Tag (PD2-H52A5) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 219 (Accession A AI13679). 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Human PD-L2, Mouse IgG1 Fc Tag (PD2-H52A5) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 219 (Accession A AI13679). 1 * 1 mg ACRBPD2-H52A5-1MG 1 * 1 mg
Hersteller-Artikelnummer:: PD2-H52A5-1MG
Lokale Artikelnummer:: ACRBPD2-H52A5-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder