Biotinylated Human PD-L2, Avitag,His Tag (recommended for biopanning) (PD2-H82E8) is expressed from human 293 cells (HEK293). It con tains AA Leu 20 - Pro 219 (Accession AAI13679). 1 * 200 G
Lieferant: ACROBIOSYSTEMS
Biotinylated Human PD-L2, Avitag,His Tag (recommended for biopanning) (PD2-H82E8) is expressed from human 293 cells (HEK293). It con tains AA Leu 20 - Pro 219 (Accession AAI13679). 1 * 200 G
PRODUKT
Erweitert / Kompakt
Biotinylated Human PD-L2, Avitag,His Tag (recommended for biopanning) (PD2-H82E8) is expressed from human 293 cells (HEK293). It con tains AA Leu 20 - Pro 219 (Accession AAI13679). 1 * 200 µG ACRBPD2-H82E8200UG 1 * 200 µG
Hersteller-Artikelnummer:: PD2-H82E8200UG
Lokale Artikelnummer:: ACRBPD2-H82E8200UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder