Biotinylated Human Prolactin, Fc,Avitag (PRN-H82F7) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Cys 227 (Acc ession Q5THQ0-1). 1 * 200 G
Lieferant: ACROBIOSYSTEMS
Biotinylated Human Prolactin, Fc,Avitag (PRN-H82F7) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Cys 227 (Acc ession Q5THQ0-1). 1 * 200 G
PRODUKT
Erweitert / Kompakt
Biotinylated Human Prolactin, Fc,Avitag (PRN-H82F7) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Cys 227 (Acc ession Q5THQ0-1). 1 * 200 µG ACRBPRN-H82F7200UG 1 * 200 µG
Hersteller-Artikelnummer:: PRN-H82F7200UG
Lokale Artikelnummer:: ACRBPRN-H82F7200UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder