Recombinat Protein,Human PSMA FOLH1 Protein, His Tag,Human PSMA, His Tag (PSA-H52H3) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession Q04609-1).This protein carries a polyhistidine tag at the N-terminus. The p a 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human PSMA FOLH1 Protein, His Tag,Human PSMA, His Tag (PSA-H52H3) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession Q04609-1).This protein carries a polyhistidine tag at the N-terminus. The p a 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human PSMA FOLH1 Protein, His Tag,Human PSMA, His Tag (PSA-H52H3) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession Q04609-1 ).This protein carries a polyhistidine tag at the N-terminus. The p a 1 * 1 mg ACRBPSA-H52H3-1MG 1 * 1 mg
Hersteller-Artikelnummer:: PSA-H52H3-1MG
Lokale Artikelnummer:: ACRBPSA-H52H3-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder