Recombinat Protein,Mouse PSMA FOLH1 Protein, His Tag,Mouse PSMA, His Tag (PSA-M5245) is expressed from human 293 cells (HEK293). It contains AA Ile 44 - Ala 752 (Accession O35409-1).This protein carries a polyhistidine tag at the N-terminus. The pr l 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Mouse PSMA FOLH1 Protein, His Tag,Mouse PSMA, His Tag (PSA-M5245) is expressed from human 293 cells (HEK293). It contains AA Ile 44 - Ala 752 (Accession O35409-1).This protein carries a polyhistidine tag at the N-terminus. The pr l 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Mouse PSMA FOLH1 Protein, His Tag,Mouse PSMA, His Tag (PSA-M5245) is expressed from human 293 cells (HEK293). It contains AA Ile 44 - Ala 752 (Accession O35409-1).This protein carries a polyhistidine tag at the N-terminus. The pr l 1 * 1 mg ACRBPSA-M5245-1MG 1 * 1 mg
Hersteller-Artikelnummer:: PSA-M5245-1MG
Lokale Artikelnummer:: ACRBPSA-M5245-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder