Recombinat Protein,Human R-Spondin 1 RSPO1 (21-146) Protein, His Tag,Human R-Spondin 1 (21-146), His Tag (RS6-H4220) is expressed f rom human 293 cells (HEK293). It contains AA Ser 21 - Ala 146 (Accession AAI14967).This protein carries a polyhistidi 1 * 50 G
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human R-Spondin 1 RSPO1 (21-146) Protein, His Tag,Human R-Spondin 1 (21-146), His Tag (RS6-H4220) is expressed f rom human 293 cells (HEK293). It contains AA Ser 21 - Ala 146 (Accession AAI14967).This protein carries a polyhistidi 1 * 50 G
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human R-Spondin 1 RSPO1 (21-146) Protein, His Tag,Human R-Spondin 1 (21-146), His Tag (RS6-H4220) is expressed f rom human 293 cells (HEK293). It contains AA Ser 21 - Ala 146 (Accession AAI14967).This protein carries a polyhistidi 1 * 50 µG ACRBRS6-H4220-50UG 1 * 50 µG
Hersteller-Artikelnummer:: RS6-H4220-50UG
Lokale Artikelnummer:: ACRBRS6-H4220-50UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder