Recombinat Protein,Human S100A10 Protein, His Tag,Human S100A10, His Tag (S10-H5127) is expressed from E.coli cells. It contains AA Met 1 - Lys 97 (Accession AAH15973).This protein carries a polyhistidine tag at the C-terminus. The protein has a cal 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human S100A10 Protein, His Tag,Human S100A10, His Tag (S10-H5127) is expressed from E.coli cells. It contains AA Met 1 - Lys 97 (Accession AAH15973).This protein carries a polyhistidine tag at the C-terminus. The protein has a cal 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human S100A10 Protein, His Tag,Human S100A10, His Tag (S10-H5127) is expressed from E.coli cells. It contains AA Met 1 - Lys 97 (Accession AAH15973).This protein carries a polyhistidine tag at the C-terminus. The protein has a cal 1 * 1 mg ACRBS10-H5127-1MG 1 * 1 mg
Hersteller-Artikelnummer:: S10-H5127-1MG
Lokale Artikelnummer:: ACRBS10-H5127-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder