Recombinat Protein,Rhesus macaque SLAMF7 CRACC CD319 Protein, His Tag,Rhesus macaque SLAMF7, His Tag (SL7-R52H7) is expressed from human 293 cells (HEK293). It contains AA Ser 23 - Met 226 (Accession F7HQ72-1).This protein carries a polyhistidine t e 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Rhesus macaque SLAMF7 CRACC CD319 Protein, His Tag,Rhesus macaque SLAMF7, His Tag (SL7-R52H7) is expressed from human 293 cells (HEK293). It contains AA Ser 23 - Met 226 (Accession F7HQ72-1).This protein carries a polyhistidine t e 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Rhesus macaque SLAMF7 CRACC CD319 Protein, His Tag,Rhesus macaque SLAMF7, His Tag (SL7-R52H7) is expressed from human 293 cells (HEK293). It contains AA Ser 23 - Met 226 (Accession F7HQ72-1).This protein carries a polyhistidine t e 1 * 1 mg ACRBSL7-R52H7-1MG 1 * 1 mg
Hersteller-Artikelnummer:: SL7-R52H7-1MG
Lokale Artikelnummer:: ACRBSL7-R52H7-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder