SARS-CoV-2 Spike Trimer His Tag (B.1.1.529/Omicron) (SPN-C52Hz) is the ectodomain of SARS-CoV-2 spike protein which contains AA Val 16 - Pro 1213 (Accession # QHD43416.1 (A67V HV69-70del T95I G142D VYY143-145del N211del L212I ins214EPE G339D S375F Y 1 * 500 G
Lieferant: ACROBIOSYSTEMS
SARS-CoV-2 Spike Trimer His Tag (B.1.1.529/Omicron) (SPN-C52Hz) is the ectodomain of SARS-CoV-2 spike protein which contains AA Val 16 - Pro 1213 (Accession # QHD43416.1 (A67V HV69-70del T95I G142D VYY143-145del N211del L212I ins214EPE G339D S375F Y 1 * 500 G
PRODUKT
Erweitert / Kompakt
SARS-CoV-2 Spike Trimer His Tag (B.1.1.529/Omicron) (SPN-C52Hz) is the ectodomain of SARS-CoV-2 spike protein which contains AA Val 16 - Pro 1213 (Accession # QHD43416.1 (A67V HV69-70del T95I G142D VYY143-145del N211del L212I ins214EPE G339D S375F Y 1 * 500 µG ACRBSPNC52HZ-500UG 1 * 500 µG
Hersteller-Artikelnummer:: SPNC52HZ-500UG
Lokale Artikelnummer:: ACRBSPNC52HZ-500UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder