Biotinylated Human TACI, Fc,Avitag (TAI-H82F6) is expressed from human 293 cells (HEK293). It contains AA Ser 2 - Thr 166 (Accession O14836-1). 1 * 200 G
Lieferant: ACROBIOSYSTEMS
Biotinylated Human TACI, Fc,Avitag (TAI-H82F6) is expressed from human 293 cells (HEK293). It contains AA Ser 2 - Thr 166 (Accession O14836-1). 1 * 200 G
PRODUKT
Erweitert / Kompakt
Biotinylated Human TACI, Fc,Avitag (TAI-H82F6) is expressed from human 293 cells (HEK293). It contains AA Ser 2 - Thr 166 (Accession O14836-1). 1 * 200 µG ACRBTAI-H82F6200UG 1 * 200 µG
Hersteller-Artikelnummer:: TAI-H82F6200UG
Lokale Artikelnummer:: ACRBTAI-H82F6200UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder