Recombinat Protein,Mouse TIM-3 HAVCR2 Protein, Fc Tag,Mouse TIM-3, Fc Tag (TI3-M5252) is expressed from human 293 cells (HEK293). I t contains AA Arg 20 - Arg 191 (Accession AAL65156.1).This protein carries a human IgG1 Fc tag at the C-terminus. Th a 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Mouse TIM-3 HAVCR2 Protein, Fc Tag,Mouse TIM-3, Fc Tag (TI3-M5252) is expressed from human 293 cells (HEK293). I t contains AA Arg 20 - Arg 191 (Accession AAL65156.1).This protein carries a human IgG1 Fc tag at the C-terminus. Th a 1 * 100 G
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Mouse TIM-3 HAVCR2 Protein, Fc Tag,Mouse TIM-3, Fc Tag (TI3-M5252) is expressed from human 293 cells (HEK293). I t contains AA Arg 20 - Arg 191 (Accession AAL65156.1).This protein carries a human IgG1 Fc tag at the C-terminus. Th a 1 * 100 µG ACRBTI3-M5252100UG 1 * 100 µG
Hersteller-Artikelnummer:: TI3-M5252100UG
Lokale Artikelnummer:: ACRBTI3-M5252100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder