Human TNFR1, Fc Tag (TN1-H5251) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Thr 211 (Accession NP001056). 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Human TNFR1, Fc Tag (TN1-H5251) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Thr 211 (Accession NP001056). 1 * 100 G
PRODUKT
Erweitert / Kompakt
Human TNFR1, Fc Tag (TN1-H5251) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Thr 211 (Accession NP001056). 1 * 100 µG ACRBTN1-H5251100UG 1 * 100 µG
Hersteller-Artikelnummer:: TN1-H5251100UG
Lokale Artikelnummer:: ACRBTN1-H5251100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder