Mouse CD40, Fc Tag (TN5-M5259) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Arg 193 (Accession P27512-1). 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Mouse CD40, Fc Tag (TN5-M5259) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Arg 193 (Accession P27512-1). 1 * 100 G
PRODUKT
Erweitert / Kompakt
Mouse CD40, Fc Tag (TN5-M5259) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Arg 193 (Accession P27512-1). 1 * 100 µG ACRBTN5-M5259100UG 1 * 100 µG
Hersteller-Artikelnummer:: TN5-M5259100UG
Lokale Artikelnummer:: ACRBTN5-M5259100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder