Human CD30, Llama IgG2b Fc Tag, low endotoxin (TN8-H5250) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 37 9 (Accession NP001234.2). 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Human CD30, Llama IgG2b Fc Tag, low endotoxin (TN8-H5250) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 37 9 (Accession NP001234.2). 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Human CD30, Llama IgG2b Fc Tag, low endotoxin (TN8-H5250) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 37 9 (Accession NP001234.2). 1 * 1 mg ACRBTN8-H5250-1MG 1 * 1 mg
Hersteller-Artikelnummer:: TN8-H5250-1MG
Lokale Artikelnummer:: ACRBTN8-H5250-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder