Recombinat Protein,Human TWEAK R TNFRSF12A Protein, Fc Tag,Human TWEAK R, Fc Tag (TNA-H526a) is expressed from human 293 cells (HEK 293). It contains AA Glu 28 - Trp 79 (Accession AAH02718).This protein carries a human IgG1 Fc tag at the N-terminus. 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human TWEAK R TNFRSF12A Protein, Fc Tag,Human TWEAK R, Fc Tag (TNA-H526a) is expressed from human 293 cells (HEK 293). It contains AA Glu 28 - Trp 79 (Accession AAH02718).This protein carries a human IgG1 Fc tag at the N-terminus. 1 * 100 G
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human TWEAK R TNFRSF12A Protein, Fc Tag,Human TWEAK R, Fc Tag (TNA-H526a) is expressed from human 293 cells (HEK 293). It contains AA Glu 28 - Trp 79 (Accession AAH02718).This protein carries a human IgG1 Fc tag at the N-terminus. 1 * 100 µG ACRBTNA-H526A100UG 1 * 100 µG
Hersteller-Artikelnummer:: TNA-H526A100UG
Lokale Artikelnummer:: ACRBTNA-H526A100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder