Recombinat Protein,Biotinylated Human TPBG 5T4 Protein, His,Avitag,Biotinylated Human TPBG, His,Avitag (TPG-H82Eb) is expressed fr om human 293 cells (HEK293). It contains AA Ser 32 - Ser 355 (Accession Q13641-1).This protein carries a polyhistidin 1 * 200 G
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Biotinylated Human TPBG 5T4 Protein, His,Avitag,Biotinylated Human TPBG, His,Avitag (TPG-H82Eb) is expressed fr om human 293 cells (HEK293). It contains AA Ser 32 - Ser 355 (Accession Q13641-1).This protein carries a polyhistidin 1 * 200 G
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Biotinylated Human TPBG 5T4 Protein, His,Avitag™,Biotinylated Human TPBG, His,Avitag (TPG-H82Eb) is expressed fr om human 293 cells (HEK293). It contains AA Ser 32 - Ser 355 (Accession Q13641-1).This protein carries a polyhistidin 1 * 200 µG ACRBTPG-H82EB200UG 1 * 200 µG
Hersteller-Artikelnummer:: TPG-H82EB200UG
Lokale Artikelnummer:: ACRBTPG-H82EB200UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder