Human TRAIL R1, His Tag (TR1-H5228) is expressed from human 293 cells (HEK293). It contains AA Ala 24 - Asn 239 (Accession NP003835 ). 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Human TRAIL R1, His Tag (TR1-H5228) is expressed from human 293 cells (HEK293). It contains AA Ala 24 - Asn 239 (Accession NP003835 ). 1 * 100 G
PRODUKT
Erweitert / Kompakt
Human TRAIL R1, His Tag (TR1-H5228) is expressed from human 293 cells (HEK293). It contains AA Ala 24 - Asn 239 (Accession NP003835 ). 1 * 100 µG ACRBTR1-H5228100UG 1 * 100 µG
Hersteller-Artikelnummer:: TR1-H5228100UG
Lokale Artikelnummer:: ACRBTR1-H5228100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder