Recombinat Protein,Cynomolgus TSLP (R127A, R130S) Protein, His Tag,Cynomolgus TSLP (R127A, R130S), His Tag (TSP-C52H4) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession A0A2K5TXV0-1 (R127A, R130S)).This protein c 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Cynomolgus TSLP (R127A, R130S) Protein, His Tag,Cynomolgus TSLP (R127A, R130S), His Tag (TSP-C52H4) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession A0A2K5TXV0-1 (R127A, R130S)).This protein c 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Cynomolgus TSLP (R127A, R130S) Protein, His Tag,Cynomolgus TSLP (R127A, R130S), His Tag (TSP-C52H4) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession A0A2K5TXV0-1 (R127A, R130S)).This protein c 1 * 1 mg ACRBTSP-C52H4-1MG 1 * 1 mg
Hersteller-Artikelnummer:: TSP-C52H4-1MG
Lokale Artikelnummer:: ACRBTSP-C52H4-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder