Recombinat Protein,Human Transthyretin Prealbumin Protein, His Tag,Human Transthyretin, His Tag (TTR-H5223) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Glu 147 (Accession NP000362).This protein carries a polyhistidine tag at s 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human Transthyretin Prealbumin Protein, His Tag,Human Transthyretin, His Tag (TTR-H5223) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Glu 147 (Accession NP000362).This protein carries a polyhistidine tag at s 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human Transthyretin Prealbumin Protein, His Tag,Human Transthyretin, His Tag (TTR-H5223) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Glu 147 (Accession NP000362).This protein carries a polyhistidine tag at s 1 * 1 mg ACRBTTR-H5223-1MG 1 * 1 mg
Hersteller-Artikelnummer:: TTR-H5223-1MG
Lokale Artikelnummer:: ACRBTTR-H5223-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder