ActiveMax Mouse VEGF120, Tag Free (VE0-M4211) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 146 (Accessio n AAB22254.1 ). 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
ActiveMax Mouse VEGF120, Tag Free (VE0-M4211) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 146 (Accessio n AAB22254.1 ). 1 * 1 mg
PRODUKT
Erweitert / Kompakt
ActiveMax® Mouse VEGF120, Tag Free (VE0-M4211) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 146 (Accessio n AAB22254.1 ). 1 * 1 mg ACRBVE0-M4211-1MG 1 * 1 mg
Hersteller-Artikelnummer:: VE0-M4211-1MG
Lokale Artikelnummer:: ACRBVE0-M4211-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder