ActiveMax Human VEGF165, Tag Free (MALS verified) (VE5-H4210) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - A rg 191 (Accession P15692-4). 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
ActiveMax Human VEGF165, Tag Free (MALS verified) (VE5-H4210) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - A rg 191 (Accession P15692-4). 1 * 1 mg
PRODUKT
Erweitert / Kompakt
ActiveMax® Human VEGF165, Tag Free (MALS verified) (VE5-H4210) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - A rg 191 (Accession P15692-4). 1 * 1 mg ACRBVE5-H4210-1MG 1 * 1 mg
Hersteller-Artikelnummer:: VE5-H4210-1MG
Lokale Artikelnummer:: ACRBVE5-H4210-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder