Biotinylated Human VEGF165, His,Avitag (VE5-H82Q0) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 191 (Acce ssion P15692-4). 1 * 25 G
Lieferant: ACROBIOSYSTEMS
Biotinylated Human VEGF165, His,Avitag (VE5-H82Q0) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 191 (Acce ssion P15692-4). 1 * 25 G
PRODUKT
Erweitert / Kompakt
Biotinylated Human VEGF165, His,Avitag (VE5-H82Q0) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 191 (Acce ssion P15692-4). 1 * 25 µG ACRBVE5-H82Q0-25UG 1 * 25 µG
Hersteller-Artikelnummer:: VE5-H82Q0-25UG
Lokale Artikelnummer:: ACRBVE5-H82Q0-25UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder