Recombinat Protein,Human VSIG2 Protein, His Tag,Human VSIG2, His Tag (VS2-H5225) is expressed from human 293 cells (HEK293). It cont ains AA Val 24 - Ala 243 (Accession AAH07313).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human VSIG2 Protein, His Tag,Human VSIG2, His Tag (VS2-H5225) is expressed from human 293 cells (HEK293). It cont ains AA Val 24 - Ala 243 (Accession AAH07313).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human VSIG2 Protein, His Tag,Human VSIG2, His Tag (VS2-H5225) is expressed from human 293 cells (HEK293). It cont ains AA Val 24 - Ala 243 (Accession AAH07313).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 1 mg ACRBVS2-H5225-1MG 1 * 1 mg
Hersteller-Artikelnummer:: VS2-H5225-1MG
Lokale Artikelnummer:: ACRBVS2-H5225-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder