Recombinat Protein,Biotinylated Human VSIG8 Protein, Fc,Avitag,Biotinylated Human VSIG8, Fc,Avitag (VS8-H82F2) is expressed from hu man 293 cells (HEK293). It contains AA Val 22 - Gly 263 (Accession NP001013683.1).This protein carries a human IgG1 - 1 * 25 G
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Biotinylated Human VSIG8 Protein, Fc,Avitag,Biotinylated Human VSIG8, Fc,Avitag (VS8-H82F2) is expressed from hu man 293 cells (HEK293). It contains AA Val 22 - Gly 263 (Accession NP001013683.1).This protein carries a human IgG1 - 1 * 25 G
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Biotinylated Human VSIG8 Protein, Fc,Avitag™,Biotinylated Human VSIG8, Fc,Avitag (VS8-H82F2) is expressed from hu man 293 cells (HEK293). It contains AA Val 22 - Gly 263 (Accession NP001013683.1).This protein carries a human IgG1 - 1 * 25 µG ACRBVS8-H82F2-25UG 1 * 25 µG
Hersteller-Artikelnummer:: VS8-H82F2-25UG
Lokale Artikelnummer:: ACRBVS8-H82F2-25UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder