Ebolavirus EBOV (subtype Zaire, strain Kikwit-95) GP, His Tag (ZE5-V5221) is expressed from human 293 cells (HEK293). It contains AA Ile 33 - Asp 637 (Accession AAQ55048.1). 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Ebolavirus EBOV (subtype Zaire, strain Kikwit-95) GP, His Tag (ZE5-V5221) is expressed from human 293 cells (HEK293). It contains AA Ile 33 - Asp 637 (Accession AAQ55048.1). 1 * 100 G
PRODUKT
Erweitert / Kompakt
Ebolavirus EBOV (subtype Zaire, strain Kikwit-95) GP, His Tag (ZE5-V5221) is expressed from human 293 cells (HEK293). It contains AA Ile 33 - Asp 637 (Accession AAQ55048.1). 1 * 100 µG ACRBZE5-V5221100UG 1 * 100 µG
Hersteller-Artikelnummer:: ZE5-V5221100UG
Lokale Artikelnummer:: ACRBZE5-V5221100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder