3-(Tetrazol-5-yl)phenylboronic acid 96%
Lieferant: ACROS ORGANICS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
Achtung
445560010
ACRO445560010EA 137 EUR InStock
ACRO445560010
3-(Tetrazol-5-yl)phenylboronic acid 96%
3-(Tetrazol-5-yl)phenylboronic acid
Formel: C₇H₇BN₄O₂
MW: 189.97 g/mol
Storage Temperature: Raumtemperatur
MDL Number: MFCD06739100
CAS-Nummer: 775351-30-9
Order Now
Spezifikation Testergebnisse
Appearance White to off-white Solid
Infrared spectrum Conforms
Additional info May contain varying amounts of anhydrides
HPLC ≥95.0 %
NMR (1H-NMR) Consistent with structure
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge