Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

GSK-J4 hydrochloride 95%

Lieferant: ACROS ORGANICS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

468001000
ACRO468001000EA 681 EUR InStock
ACRO468001000
GSK-J4 hydrochloride 95%
GSK-J4 hydrochloride
Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.

Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies


Formel: C₂₄H₂₇N₅O₂∙HCl
Storage Temperature: Gefrierschrank
MDL Number: MFCD26142638
CAS-Nummer: 1797983-09-5
Order Now
Spezifikation Testergebnisse
Appearance Yellow Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge