Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Spebrutinib 95%

Lieferant: ACROS ORGANICS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

468061000
ACRO468061000EA 289 EUR InStock
ACRO468061000
Spebrutinib 95%
Spebrutinib
Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.

Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies


Formel: C₂₂H₂₂FN₅O₃
Storage Temperature: Gefrierschrank
MDL Number: MFCD25976876
CAS-Nummer: 1202757-89-8
Order Now
Spezifikation Testergebnisse
Appearance White to light brown Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure

 

PRODUKT

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge