Zinc Standard Solution, 10000 mg/L Zn in dil. Nitric Acid Standard for ICP-MS
Lieferant: ACTIVE MOTIF
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
M169.5NP.L1
ACSDM169.5NP.L1EA 231 EUR InStock
ACSDM169.5NP.L1
Zinc Standard Solution, 10000 mg/L Zn in dil. Nitric Acid Standard for ICP-MS
Standards Kationischer Standards Zink-Standards
Mg 10000 mg/L in diluted nitric acid for ICP/MS.
Shelf life on ship date: 36 months - store under normal laboratory conditions, at temperatures between 15 to 25 °C.
Zertifizierungen: ISO 17034, ISO 17025, ISO 9001.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge