Liquid Viewer, Apollo II
Lieferant: ADELPHI MANUFACTURING
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
6031522
ADLM6031522EA 0 EUR InStock
ADLM6031522
Liquid Viewer, Apollo II
Beleuchtung Leuchtkästen
Designed for use in pharmaceutical and laboratory environments, the Apollo II Liquid Viewer for transparent glass containers provides a simple method for the detection of visible particles in ampoules, vials, and bottles.
  • Height and angle adjustable
  • Intuitive assembly; no tools required
  • Light level between 2,000 and 3,750 Lux
  • Unit can fold for convenient storage, with the inspection surfaces folding together to reduce chance of damage
The unit is packaged in sustainably sourced foam, made from polyethylene resin which is derived from renewable sugarcane-based feedstock.
The Apollo II Liquid Viewer is suitable for transparent glass containers. Adelphi Manufacturing also offer the Apollo II High Intensity Liquid Viewer for coloured glass containers, and the Apollo I Liquid Viewer, with additional magnifier.
Zertifizierungen: This unit conforms to the European Pharmacopoeia Specification 2.9.20 and to the US Pharmacopoeia – USP chapter 41 (1790), section 6.1.
Adelphi Manufacturing’s ISO 9001:2015 certification guarantees the high quality finish of this product. Supporting IQ/OQ documentation can be supplied. User manuals are available in English, Spanish, French, German, Dutch and Czech – additional languages are also available upon request.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge