Spare LED tubes for Apollo II and Apollo II high intensity liquid viewers
Lieferant: ADELPHI MANUFACTURING
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
6031535
ADLM6031535EA 0 EUR InStock
ADLM6031535
Spare LED tubes for Apollo II and Apollo II high intensity liquid viewers
Beleuchtung LED-Beleuchtung
Pack of two 10 Watt LED bulbs (550 mm long) with a diffuser, which give a light level of between 2000 and 3750 Lux. The benefit of using LED tubes is more consistent light over the years compared with fluorescent tubes. These LED bulbs have a service life of 50,000 hours.
  • Colour temperature: 4000 K
  • Colour Rendering Index (CRI) Ra: greater than 80
  • Luminous flux at 25 °C: 950 - 1150 Im
The same bulbs to be used for Apollo II (for transparent glass) as well as Apollo II High Intensity (for colour glass).
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge