Membrane filters, hydrophobic, PTFE
Lieferant: ADVANTEC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
T020A142C T020A293D
ADVTT020A142CEA 885 EUR InStock
ADVTT020A142C ADVTT020A293D
Membrane filters, hydrophobic, PTFE
Filter Membranfilter
Hydrophobic PTFE membrane filters behave as an absolute retentive membrane.
  • Thin and highly porous
  • Inert to most agressive solvents, strong acids and bases
  • Unsupported
Used to sterilise gases, vent gas from aqueous solutions, or sterilise or clarify aggressive chemicals.
Operating temperature range: –120 to 260 °C.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge