Tracheal retractors, Aesculap®
Lieferant: AESCULAP AG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BT106R BT107R BT103R
AESCBT106REA 0 EUR InStock
AESCBT106R AESCBT107R AESCBT103R
Tracheal retractors, Aesculap®
Retraktoren
These handheld retractors are used during tracheotomy procedures. They are available as a single or multi-prong instrument where the prong(s) hold back cricoid cartilage prior to the tracheal incision.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge