Instrument boxes
Lieferant: AGENCINOX
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
8036B 8036R 8036J 8029 8030 8031 8034 8036
AGEN8036BEA 58.9 EUR InStock
AGEN8036B AGEN8036R AGEN8036J AGEN8029 AGEN8030 AGEN8031 AGEN8034 AGEN8036
Instrument boxes
Schalen/Schüsseln/Töpfe Instrumentenschalen
Aluminium, anodised.
  • Smooth surfaces for fast, easy cleaning
  • Coloured
  • Good chemical resistance, very good thermal resistance
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge