Deoxynucleotide mix
Lieferant: AGILENT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
200415
AGLS200415EA 279 EUR InStock
AGLS200415
Deoxynucleotide mix
Nukleinsäure-Reagenzien Nukleotide
The PCR grade deoxynucleotide mix includes 25 mM of each standard dNTP (GATC) for use with any PCR application, including real-time quantitative PCR.
  • Ideal for optimal PCR performance
  • Produces high-quality reaction products
  • Withstands multiple freeze-thaw without compromising efficiency
Lieferumfang: Includes 400 µl of 100 mM dNTP mix (25 mM of each dNTP), adequate for 1000 standard 50 µl primer-extension reactions or 2000 reactions of 25 µl.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge